Vanadium carbide, 99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1214114

Quantity 25g

 View more versions of this product

Catalog No. AA1214114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12070-10-9
    Physical Form Powder
    Assay Percent Range 99% metal basis
    Molecular Formula VC (VC0?88 by X-Ray)
    Formula Weight 62.95
    MDL Number MFCD00049698

    Specifications

    Formula Weight 62.95
    Quantity 25g
    Color Black
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99% metal basis
    Molecular Formula VC (VC0?88 by X-Ray)
    MDL Number MFCD00049698
    CAS 12070-10-9

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit