Alfa Aesar™ Vanadium gallide, 99% (metals basis)

Manufacturer: Alfa Aesar™ 1751906

Quantity 5g

 View more versions of this product

Catalog No. AA1751906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12024-15-6
    Formula Weight 222.55g/mol
    MDL Number MFCD02091753
    UN Number UN3285
    Chemical Name or Material Vanadium gallide

    Specifications

    Mesh Size −100 Mesh
    Melting Point 1720°C
    Quantity 5g
    CAS 12024-15-6
    Physical Form Powder
    Formula Weight 222.55g/mol
    MDL Number MFCD02091753
    UN Number UN3285
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
    Chemical Name or Material Vanadium gallide
    Assay (metals basis)

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit