Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium Granules, 1-3mm (0.04-0.12 in.), 99.7% (Metals basis), Alfa Aesar™

Click to view available options
Quantity:
10g
50g
250g
7440-62-2
V
50.941
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde
23990
CHEBI:35170
vanadium
[V]
7440-62-2
V
50.941
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde
23990
CHEBI:35170
vanadium
[V]

For Vacuum Deposition

Quantity 250g
Appearance Light Gray granules
Density 6.11g/cm3
Assay Percent Range 99.7% (Metals basis)
Odor Odorless
Physical Form Granular
Grade Vacuum Deposition
Color Gray
Chemical Name or Material Vanadium

EINECSNumber : 231-171-1

TSCA : Yes

Recommended Storage : Ambient temperatures