Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Vanadium(IV) chloride, 99+%, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 8768535

Quantity 450g

 View more versions of this product

Catalog No. AA8768535

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7632-51-1
192.75
UN2444
Vanadium (IV) chloride
VCl4
MFCD00061343
Vanadium tetrachloride

Specifications

7632-51-1
VCl4
MFCD00061343
Vanadium tetrachloride
Cylinder with ball valve
148.5°C (755mmHg)
450g
≥99%, (metals basis)
Liquid
192.75
UN2444
1.816g/mL at 30°C
Moisture Sensitive
−28°C
Vanadium (IV) chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: VANADIUM TETRACHLORIDE

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit