Alfa Aesar™ Vanadium(IV) fluoride, 95%

Manufacturer: Alfa Aesar™ 1154306

Quantity 5g

 View more versions of this product

Catalog No. AA1154306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10049-16-8
    Linear Formula VF4
    Formula Weight 126.94
    MDL Number MFCD00042554
    UN Number UN2923
    Synonym Vanadium tetrafluoride
    Chemical Name or Material Vanadium(IV) fluoride

    Specifications

    Formula Weight 126.94
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00042554
    UN Number UN2923
    Health Hazard 2 GHS H Statement
    H301-H331-H314-H318
    Toxic if swallowed.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Physical Form Powder
    Solubility Information Soluble in water (blue color), acetone (deep green color), and glacial acetic acid (blue-green color)
    Health Hazard 1 Danger
    Density 2.975g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Sensitivity Moisture Sensitive
    Linear Formula VF4
    CAS 10049-16-8
    Synonym Vanadium tetrafluoride
    Color Yellow to brown
    Melting Point 325°C (dec.)
    Quantity 5g
    Chemical Name or Material Vanadium(IV) fluoride
    Assay 95%

    GHS H Statement
    H301-H331-H314-H318
    Toxic if swallowed.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit