Alfa Aesar™ Vanadium(IV) oxide bis(2,4-pentanedionate)

As a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment

Manufacturer: Alfa Aesar™ 8111930

Quantity 250g

 View more versions of this product

Catalog No. AA8111930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3153-26-2
    Molecular Formula C10H14O5V
    Formula Weight 265.16
    MDL Number MFCD00000032
    UN Number UN3285
    Synonym Vanadyl acetylacetonate; Vanadyl 2,4-pentanedionate
    Chemical Name or Material Vanadium (IV) oxide bis (2,4-pentanedionate)

    Specifications

    Formula Weight 265.16
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Powder
    Synonym Vanadyl acetylacetonate; Vanadyl 2,4-pentanedionate
    Boiling Point 140°C (13mmHg)
    Solubility Information Moderately soluble in acetone, ether and chloroform; Soluble in ethanol and benzene
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 1.4g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
    Molecular Formula C10H14O5V
    CAS 3153-26-2
    UN Number UN3285
    Melting Point 256° to 259°C
    MDL Number MFCD00000032
    Quantity 250g
    Chemical Name or Material Vanadium (IV) oxide bis (2,4-pentanedionate)

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit