Alfa Aesar™ Vanadium(IV) oxide meso-tetraphenylporphine

Manufacturer: Alfa Aesar™ 39583MC

Quantity 100mg

 View more versions of this product

Catalog No. AA39583MC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14705-63-6
    Formula Weight 679.67
    MDL Number MFCD00012154
    Synonym Tetraphenylporphine vanadium(IV) oxide
    Chemical Name or Material Vanadium(IV) oxide meso-tetraphenylporphine

    Specifications

    Formula Weight 679.67
    Odor Odorless
    Physical Form Crystalline
    MDL Number MFCD00012154
    CAS 14705-63-6
    Synonym Tetraphenylporphine vanadium(IV) oxide
    Color Purple
    Quantity 100mg
    Chemical Name or Material Vanadium(IV) oxide meso-tetraphenylporphine

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit