Vanadium(IV) Sulfate Oxide Hydrate, 99.9% (Metal Basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1109030

Quantity 250g

 View more versions of this product

Catalog No. AA1109030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 123334-20-3
    Physical Form Lumps
    Assay Percent Range 99.9%, metal basis
    Molecular Formula VOSO4·xH2O
    Formula Weight 163
    MDL Number MFCD00149933
    Synonym Vanadium oxysulfate hydrate
    Chemical Name or Material Vanadium(IV) Sulfate Oxide Hydrate

    Specifications

    Formula Weight 163
    Quantity 250g
    DOT Information DOT Class: 6.1
    EINECS Number 248-652-7
    Physical Form Lumps
    Assay Percent Range 99.9%, metal basis
    Molecular Formula VOSO4·xH2O
    MDL Number MFCD00149933
    CAS 123334-20-3
    Synonym Vanadium oxysulfate hydrate
    Health Hazard 1 Harmful if swallowed.
    Health Hazard 2 Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste.
    Chemical Name or Material Vanadium(IV) Sulfate Oxide Hydrate

    Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste.

    Harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit