Learn More
Vanadium(IV) sulfate oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Vanadium(IV) sulfate oxide hydrate is used as a catalyst for the oxidative aromatization of cyclohexenone. It is also used as a component in some food supplements. It is one of the intermediate during the extraction of vanadium from petroleum residues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Lump |
Quantity | 10 g |
Assay Percent Range | 99.9%, metal basis |
Solubility Information | Soluble in water. |
IUPAC Name | oxovanadiumbis(ylium) sulfate |
Formula Weight | 163.00 (Anhydrous) |
Density | 2.5 |
Chemical Name or Material | Vanadium(IV) Sulfate Oxide Hydrate |
Safety and Handling
Hazard Category | H302 |
Hazard Statement | Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste. |
Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
DOTInformation | DOT Class: 6.1; Packaging Group: II |
EINECSNumber | 248-652-7 |
RTECSNumber | YW1840000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.