Alfa Aesar™ Vanadium(IV,V) oxide, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 2295609

Quantity 10g

 View more versions of this product

Catalog No. AA2295609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12037-42-2
    Formula Weight 513.64
    MDL Number MFCD00064848
    UN Number UN3285
    Chemical Name or Material Vanadium(IV,V) oxide

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 513.64
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H332-H319-H335
    Harmful if swallowed.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00064848
    UN Number UN3285
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
    Odor Odorless
    CAS 12037-42-2
    Color Blue-black
    Melting Point ≈1500°C
    Quantity 10g
    Density 3.91g/mL
    Chemical Name or Material Vanadium(IV,V) oxide
    Assay 99.5%, (metals basis)

    GHS H Statement
    H302-H332-H319-H335
    Harmful if swallowed.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit