Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Vanadium naphthenate oxide, 35% in naphthenic acid (V 2.8-3.2%)

Manufacturer:  Alfa Aesar™ 3958536

Quantity 500g

 View more versions of this product

Catalog No. AA3958536


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

68553-60-6
MFCD00148763
UN2810
Vanadium naphthenate oxide

Specifications

68553-60-6
MFCD00148763
35% in naphthenic acid (V 2.8 to 3.2%)
500g
Liquid
UN2810
1.04g/mL
Vanadium naphthenate oxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H330-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit