Alfa Aesar™ Vanadium naphthenate oxide, 35% in naphthenic acid (V 2.8-3.2%)

Manufacturer: Alfa Aesar™ 3958522

Quantity 100g

 View more versions of this product

Catalog No. AA3958522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 68553-60-6
    MDL Number MFCD00148763
    UN Number UN2810
    Chemical Name or Material Vanadium naphthenate oxide

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    CAS 68553-60-6
    Quantity 100g
    Health Hazard 1 Danger
    TSCA TSCA
    MDL Number MFCD00148763
    UN Number UN2810
    Health Hazard 2 GHS H Statement
    H300-H330-H315-H319-H335
    Fatal if swallowed.
    Fatal if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 1.04g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Concentration 35% in naphthenic acid (V 2.8 to 3.2%)
    Chemical Name or Material Vanadium naphthenate oxide

    GHS H Statement
    H300-H330-H315-H319-H335
    Fatal if swallowed.
    Fatal if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit