Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanadium Turnings, 2.5cm (1.0 in.) & down, 99.7% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1042018

Quantity 50g

 View more versions of this product

Catalog No. AA1042018

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-62-2
99.7% (Metals basis)
Vanadium
Turnings, 2.5cm (1.0 in.) and down
MFCD00011453

Specifications

50g
Silver Gray turnings
99.7% (Metals basis)
Odorless
Vanadium
7440-62-2
Turnings, 2.5cm (1.0 in.) and down
MFCD00011453
Silver-gray
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit