Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanadium(V) oxide, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1750.0°C |
Melting Point | 690.0°C |
Color | Blue |
Physical Form | Powder, Crystals and/or Chunks |
Quantity | 100 g |
Assay Percent Range | 98% min |
Linear Formula | V2O5 |
Merck Index | 15, 10107 |
Solubility Information | Solubility in water: 8g/L. Other solubilities: soluble in acids and alkalies,insoluble in alcohol |
IUPAC Name | [(dioxovanadio)oxy]dioxovanadium |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Suspected of damaging the unborn child. Suspected of causing genetic defects. Harmful if swallowed. Harmful if inhaled. May cause respiratory irritation. Causes damage to organs through prolonged or re |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. |
EINECSNumber | 215-239-8 |
RTECSNumber | YW2450000 |
TSCA | TSCA |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction