Alfa Aesar™ Vanadium(V) triisopropoxide oxide, 96%

Manufacturer: Alfa Aesar™ 8979806

Quantity 5g

 View more versions of this product

Catalog No. AA8979806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5588-84-1
    Molecular Formula C9H21O4V
    Linear Formula VO[CHO(CH3)2]3
    Formula Weight 242.2
    MDL Number MFCD00015017
    UN Number UN1993
    Synonym Isopropyl orthovanadate; Vanadium(V) oxytriisopropoxide
    Chemical Name or Material Vanadium (V) triisopropoxide oxide

    Specifications

    Formula Weight 242.2
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    MDL Number MFCD00015017
    CAS 5588-84-1
    Boiling Point 81° to 83°C (2mmHg)
    Flash Point 45°C (113°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C9H21O4V
    UN Number UN1993
    TSCA TSCA
    Physical Form Liquid
    Sensitivity Moisture Sensitive
    Synonym Isopropyl orthovanadate; Vanadium(V) oxytriisopropoxide
    Color Pale yellow-orange
    Melting Point −12°C
    Quantity 5g
    Linear Formula VO[CHO(CH3)2]3
    Density 1.04g/mL
    Chemical Name or Material Vanadium (V) triisopropoxide oxide
    Assay 96%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit