Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanado - Molybdate Reagent, AOAC, Ricca Chemical
$168.00 - $516.00
Specifications
Chemical Name or Material | Vanado - Molybdate Reagent |
---|---|
Color | Colorless to Yellow |
CAS Min % | 70.0, 26.29, 1.41, 0.07 |
CAS Max % | 73.0, 27.37, 1.46, 0.07 |
CAS | 7601-90-3, 12054-85-2, 7803-55-6 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
898516
|
Ricca Chemical Company
898516 |
500 mL | Natural Poly Bottle |
Each for $168.00
|
|
||||
898532
|
Ricca Chemical Company
898532 |
1 L | Natural Poly Bottle |
Each for $230.00
|
|
||||
89851
|
Ricca Chemical Company
89851 |
4 L | Natural Poly Bottle |
Each for $516.00
|
|
||||
Specifications
Vanado - Molybdate Reagent | |
70.0, 26.29, 1.41, 0.07 | |
7601-90-3, 12054-85-2, 7803-55-6 | |
dihydrogen oxide, dihydrogen monoxide | |
Natural Poly Bottle |
Colorless to Yellow | |
73.0, 27.37, 1.46, 0.07 | |
Liquid | |
AOAC |
Safety and Handling
ShelfLife : 24 Months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title