Learn More
Cole-Parmer Davis Instruments 6153 Wireless Weather Station with 24-Hour Fan Aspirated Radiation Shield  DFS Item
Manufacturer:  Cole-Parmer 3454265
This powerful station transmits data up to 1000 ft or even further with an optional wireless repeater. Measured parameters update every 2.5 seconds as compared to 3 to 15 minutes found on other stations.
Catalog No. 50-628-420