Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAP-A Mouse anti-Human, Biotin, Clone: OTI10E10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274825B

Catalog No. NBP274825B


Add to cart

Description

Description

VAP-A Monoclonal antibody specifically detects VAP-A in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

VAP-A
OTI10E10
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
VAPA
Recombinant protein fragment corresponding to amino acids 2-227 of human VAP-A (NP_919415) produced in E.coli.
Immunogen affinity purified
Signal Transduction
Primary
9218.0
Immunohistochemistry, Western Blot
Biotin
PBS with 0.05% Sodium Azide
hVAP-33, MGC3745,33 kDa VAMP-associated protein, VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa, VAMP-A, VAP-33, VAP33VAMP (vesicle-associated membrane protein)-associated protein A (33kD), VAP-AVAMP-associated protein A, vesicle-associated membrane protein-associated protein A
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit