Learn More
VAPA Mouse anti-Human, Clone: OTI10E10, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809100
Catalog No. 50-166-9268
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a type IV membrane protein. It is present in the plasma membrane and intracellular vesicles. It may also be associated with the cytoskeleton. This protein may function in vesicle trafficking, membrane fusion, protein complex assembly and cell motility. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
Specifications
VAPA
Monoclonal
Unconjugated
VAPA
Lyophilized
VAPA
Human recombit protein fragment corresponding to amino acids 2-227 of human VAPA produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI10E10
PBS with 8% trehalose and no preservative; pH 7.3
Q9P0L0
VAP-33, VAP-A, VAP33, hVAP-33
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9218