Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAPA Mouse anti-Human, Clone: OTI10E10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527092

Catalog No. PIMA527092


Add to cart

Description

Description

The protein encoded by this gene is a type IV membrane protein. It is present in the plasma membrane and intracellular vesicles. It may also be associated with the cytoskeleton. This protein may function in vesicle trafficking, membrane fusion, protein complex assembly and cell motility. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications

Specifications

VAPA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9P0L0
VAP-33, VAP-A, VAP33, hVAP-33, 33 kDa VAMP-associated protein, VAMP-A, VAMP-associated protein A, vesicle-associated membrane protein-associated protein A
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9218
Immunohistochemistry (Paraffin), Western Blot
OTI10E10
Unconjugated
VAPA
Liquid
VAPA
Human recombit protein fragment corresponding to amino acids 2-227 of VAPA produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit