Learn More
Cole-Parmer™ VapLock™ Activated Media Exhaust Filters
Catalog No. 13311186 Shop All Oakton Products
Change view
Click to view available options
Includes:
300 g Activated Carbon
75 g Activated Carbon
75 g Activated Carbon, Breakthrough Indicator
Quantity:
1 Ea.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Quantity
13-311-186 300 g Activated Carbon 1 Ea.
13-311-181 75 g Activated Carbon 1 Ea.
13-311-182 75 g Activated Carbon, Breakthrough Indicator 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-311-186 Supplier Cole-Parmer™ Supplier No. 1201984
Add to Cart
Add to Cart
VapLock™ Activated Media Exhaust Filters reduce harmful chemical emissions and odors from solvent containers
Use with indicators to set a replacement schedule
Warranty and Services
None
Specifications
For Use With (Equipment) VapLock Solvent Containment Systems
Includes 300 g Activated Carbon
Material Activated Carbon
Thread Size 1/4 in. NPT(F)
For Use With (Application) Solvent Safety
Quantity 1 Ea.
Product Type Exhaust Filter