Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Activated Media Exhaust Filters

VapLock™ Activated Media Exhaust Filters reduce harmful chemical emissions and odors from solvent containers

$76.40 - $233.00

Specifications

For Use With (Equipment) VapLock Solvent Containment Systems
For Use With (Application) Solvent Safety
Thread Size (English) 1/4 in. NPT(F)
View More Specs

Products 4
Catalog Number Type Material Price Quantity & Availability  
Catalog Number Type Material Price Quantity & Availability  
13-311-181
View Documents
Exhaust Filter, 75 g activated carbon, 1/4 in. NPT(F); 1/ea Activated Carbon
Each for $76.40
Add to cart
 
13-311-182
View Documents
Exhaust Filter with Indicator, 75 g activated carbon, 1/4 in. NPT(F); 1/ea Activated Carbon with Breakthrough Indicator
Each for $160.80
Add to cart
 
13-311-186
View Documents
Exhaust Filter, 300 g activated carbon, 1/4 in. NPT(F); 1/ea Activated Carbon
Each for $233.00
Add to cart
 
13-311-183
View Documents
Exhaust Filter, 75 g activated carbon, 1/4 in. NPT(F); 5/Pk. Activated Carbon This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Use with indicators to set a replacement schedule

Warranty and Services

None

Specifications

Specifications

VapLock Solvent Containment Systems
Solvent Safety
1/4 in. NPT(F)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit