Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, NPT(M)
VapLock™ Adapters for Exhaust Filters, NPT(M) feature various adapter configurations allowing quick and easy connection to emission sources
 View more versions of this product
Catalog No. 13-311-196
Add to cart
Description
Description
  • Fit VapLock exhaust filters, which have 1/4-inch NPT(F) ports
  • May require additional adapters to fit your container
Warranty and Services
None
Specifications
Specifications
Filter Adapter; 1/ea
VapLock Exhaust Filters
90 elbow, 1/4 in. NPT(M) x 1/4 in. NPT(M)
Solvent Safety
Gray Polypropylene
1/4 in. NPT(M) x 1/4 in. NPT(M)