Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, NPT(M)
VapLock™ Adapters for Exhaust Filters, NPT(M) feature various adapter configurations allowing quick and easy connection to emission sources
$2.18 - $20.75
Specifications
Type | Filter Adapter; 1/ea |
---|---|
For Use With (Application) | Solvent Safety |
For Use With (Equipment) | VapLock Exhaust Filters |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
13-311-188
|
Cole-Parmer™
1201986 |
1/4 in. NPT(M) x 1/4 in. NPT(M) |
Each for $2.18
|
|
|||||
13-311-189
|
Cole-Parmer™
1201987 |
1/4 in. NPT(M) x 1/2 in. NPT(M) |
Each for $3.32
|
|
|||||
13-311-191
|
Cole-Parmer™
1201989 |
90 elbow, 1/4 in. NPT(M) x 1/2 in. NPT(M) |
Each for $20.75
|
|
|||||
13-311-195
|
Cole-Parmer™
1201993 |
45 elbow, 1/4 in. NPT(M) x 1/4 in. NPT(M) |
Each for $16.85
|
|
|||||
13-311-196
|
Cole-Parmer™
1201994 |
90 elbow, 1/4 in. NPT(M) x 1/4 in. NPT(M) |
Each for $18.41
|
|
|||||
13-311-202
|
Cole-Parmer™
1202000 |
1/8 in. NPT(M) x 1/4 in. NPT(M) |
Each for $3.21
|
|
|||||
Description
- Fit VapLock exhaust filters, which have 1/4-inch NPT(F) ports
- May require additional adapters to fit your container
Warranty and Services
None
Specifications
Filter Adapter; 1/ea | |
Solvent Safety | |
VapLock Exhaust Filters |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title