Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system
 View more versions of this product
Catalog No. 13-311-108
Add to cart
Description
Description
  • Select from a variety of cap sizes to fit your container
  • Different materials, numbers and sizes of ports help customize to your application
Warranty and Services
None
Specifications
Specifications
Solvent Safety
Polypropylene
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Solvent-Compatible Containers
60 mm
Solvent Waste Manifold, PP, 60 mm, 1 x 1/2 in. NPTF, 3 x 1/4 in. NPTF, 6 x 1/4 in.-28 UNFF; 1/ea