Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ Vaporizer Service Exchange Iso Tec 3EX, 250 mL capacity (Specify Fill Type, Mount)
SDP

Maintains device accuracy, proper function and prevents exposure from leaks. A fully serviced and calibrated vaporizer of the same type is shipped to the customer. Customer uses prepaid return label to send their vaporizer back to our service center. No downtime.

Supplier:  SOMNI Scientific™ VX6029

Catalog No. 04-777-162


Add to Cart

Description

Description

  • Isoflurane, 3EX extended capacity (250ml)
  • Specify Fill type, Mount and Serial number when ordering
  • Maintains accuracy
  • Prevents exposure from leaks
  • Full Service and Calibration
    • Certificate of Calibration included
  • Service interval based on manufacturer recommendation
  • Warranty provided
Specifications

Specifications

RM-17003, RM-17004, AMD-17004, TT-17000, OC-21003, OC-21004
Brass and Alloys
13.5 cm
TEC 3EX IsoFlurane (250 mL)
14 cm
14 cm
Animal Inhalant anesthesia
Mounting hardware, Calibration Certificate
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Veterinary Use Only