Learn More
Justrite™ VaporTrap™ Polypropylene, 83mm Carboy Caps with Filter Kit, Four Ports 1/8 in. OD Tubing, Four Ports 1/4 in. OD Tubing
For Use With: carboys with 83mm cap
Supplier:  Justrite™ 12829
Catalog No. 19-830-389
Add to Cart
Description
Description
  • Carbon filter prevents volatile organic compound vapors
Specifications
Specifications
Four (4) 1/8 in. Tubing OD Ports and Four (4) 1/4 in. Tubing OD Ports
Polypropylene
carboys with 83mm cap
5 x 5.5 x 11 in.
Caps