Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ VaporTrap™ Polypropylene, 83mm Carboy Caps with Filter Kit, Six Ports 1/8'' OD Tubing

For Use With: carboys with 83mm cap

Supplier:  Justrite™ 12825

Catalog No. 19-830-385


Add to Cart
This item is not returnable. View return policy

Description

Description

  • Carbon filter prevents volatile organic compound vapors
Specifications

Specifications

Six (6) 1/8 in. Tubing OD Ports
Polypropylene
carboys with 83mm cap
5 x 5.5 x 11 in.
Caps
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit