Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ VaporTrap™ HDPE, UN/DOT-Certified Carboy with Filter Kit, Four Ports 1/8 in. and Four Ports 1/4 in.
For Use With: Safe transport of hazardous liquids; Closure Material: Polypropylene; Color: White
Supplier: Justrite™ 12845
Description
- For packaging group II and hazardous chemicals
- Carbon filter prevents volatile organic compound vapors
- Two-piece cap design
Specifications
5.3 gal. | |
Polypropylene | |
Four (4) 1/8 in. Tubing OD Ports and Four (4) 1/4 in. Tubing OD Ports | |
287 x 235 x 445 mm | |
UN/DOT Carboy with Filter Kit | |
17.5 in. | |
Rectangular | |
White |
UN/DOT | |
70 mm | |
20 L | |
HDPE | |
Safe transport of hazardous liquids | |
11.3 in. | |
9.25 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title