Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ VaporTrap™ HDPE, UN/DOT-Certified Carboy with Filter Kit, Four Ports 1/8 in. and Four Ports 1/4 in.
For Use With: Safe transport of hazardous liquids; Closure Material: Polypropylene; Color: White
$508.20 - $542.85
Specifications
Certifications/Compliance | UN/DOT |
---|---|
Closure Material | Polypropylene |
Closure Size | 70 mm |
Description | Four (4) 1/8 in. Tubing OD Ports and Four (4) 1/4 in. Tubing OD Ports |
Material | HDPE |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
19-830-398
|
Justrite™
12844 |
13.5 L |
Each for $508.20
|
|
19-830-399
|
Justrite™
12845 |
20 L |
Each for $542.85
|
|
Description
- For packaging group II and hazardous chemicals
- Carbon filter prevents volatile organic compound vapors
- Two-piece cap design
Specifications
UN/DOT | |
70 mm | |
HDPE | |
Safe transport of hazardous liquids | |
Rectangular | |
White |
Polypropylene | |
Four (4) 1/8 in. Tubing OD Ports and Four (4) 1/4 in. Tubing OD Ports | |
UN/DOT Carboy with Filter Kit | |
11.3 in. | |
9.25 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title