Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ VaporTrap™ HDPE, UN/DOT-Certified Carboy with Filter Kit and Six Ports 1/8 in.

For Use With: Safe transport of hazardous liquids; Closure Material: Polypropylene; Color: White

Supplier:  Justrite™ 12839

 View more versions of this product

Catalog No. 19-830-393


Add to Cart

Description

Description

  • For packaging group II and hazardous chemicals
  • Carbon filter prevents volatile organic compound vapors
  • Two-piece cap design
Specifications

Specifications

5.3 gal.
Polypropylene
Six (6) 1/8 in. Tubing OD Ports
287 x 235 x 445 mm
UN/DOT Carboy with Filter Kit
17.5 in.
Rectangular
White
UN/DOT
70 mm
20 L
HDPE
Safe transport of hazardous liquids
11.3 in.
9.25 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit