Learn More
Justrite™ VaporTrap™ HDPE, Round Carboy with Filter Kit, Four Ports 1/8 in. Tubing, Three Ports 1/4 in., and One Port 1/4 in. or 3/8 in. Hose Barb
For Use With: Designed to collect waste from one or more HPLC machines; Closure Material: Polypropylene; Color: White
Supplier:  Justrite™ 12824
Catalog No. 19-830-384
Add to Cart
Description
Description
  • Carbon filter prevents volatile organic compound vapors
  • Two-piece cap design eliminates need to disconnect tubing from adapter to install closed cap
Specifications
Specifications
5.3 gal.
83 mm
11 in.
HDPE
Designed to collect waste from one or more HPLC machines
23 in.
Cylindrical
Polypropylene
Four (4) 1/8 in. Tubing OD Ports, Three (3) 1/4 in. Tubing OD Ports and One (1) 1/4 or 3/8 in. Tubing ID Hose Barb Ports
20 L
Carboy with Filter Kit
23 in.
Closure
White