Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix Varicella-Zoster Virus Culture Fluid
Infectious virus preparation designed to meet research and product development needs
Specifications
Biosafety Level | 2 |
---|---|
Certifications/Compliance | ISO, 13485 |
Content And Storage | -65°C |
Form | Solution |
Media Type | Vial |
Alert:
Contains live and infectious organisms.
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
22-157-712
|
ZeptoMetrix
0810169CF |
Microbiology Media |
Each for $1,795.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-711
|
ZeptoMetrix
0810168CF |
Microbiology Media |
Each for $1,795.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-713
|
ZeptoMetrix
0810170CF |
Microbiology Media |
Each for $1,795.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-714
|
ZeptoMetrix
0810171CF |
Microbiology Media |
Each for $1,795.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-710
|
ZeptoMetrix
0810167CF |
Microbiology Media |
Each for $1,795.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-715
|
ZeptoMetrix
0810172CF |
Microbiology Media |
Each for $1,795.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-716
|
ZeptoMetrix
0810173CF |
Microbiology Media |
Each for $1,795.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-717
|
ZeptoMetrix
0810175CF |
Microbiology Media |
Each for $1,795.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Nucleic acid/molecular testing, limit of detection (LOD) studies, cross-reactivity studies and other viral-based assays
Specifications
2 | |
-65°C | |
Vial | |
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
ISO, 13485 | |
Solution | |
Microbiology Media | |
1 mL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title