Learn More
Varicella zoster virus MCP Mouse anti-Virus, Clone: VA035, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13666
Catalog No. 89-033-629
Add to cart
Description
Description
Mouse monoclonal antibody raised against varicella zoster virus (VZV) MCP.
  • High specificity
  • High reactivity
Specifications
Specifications
Varicella zoster virus MCP
VA035
Mouse monoclonal antibody raised against varicella zoster virus (VZV) MCP.
In PBS, pH 7.4.
Mouse
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus
Immunofluorescence
Unconjugated
Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
IgG1, kappa
RUO
Primary
This antibody reacts with MCP of varicella zoster virus (VZV).
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)