Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Varistain™ Gemini ES Automated Slide Stainer

Maximize efficiency by organizing staining workflow with the multidimensional Thermo Scientific™ Varistain™ Gemini ES Automated Slide Stainer.

Specifications

Certifications/Compliance Designed to meet IEC 61010. CSA approved. UL listed. CE marked (IVDD compliant).
Dimensions (L x W x H) 28 x 28.5 x 34.5 in. (71 x 72 x 88cm)
Height (English) 34.5 in.
Height (Metric) 88cm
Length (English) 28 in.
View More Specs

Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
A78000004
View Documents
Thermo Scientific™
A78000004
Without heat, with monitor This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description

Description

Maximize efficiency by organizing staining workflow with the multidimensional Thermo Scientific™ Varistain™ Gemini ES Automated Slide Stainer. The Gemini ES is an open system that utilizes Thermo Scientific stains and reagents. This stainer's single button, easy-to-use software schedules runs, allocates reagents, optimizes reagent layout, and calculates the most efficient route for each protocol.

Performance

  • Loads 20-slide baskets with up to 100 slides at once
  • Step-start available from any station
  • Urgent Start gives user opportunity to prioritize “urgent” baskets
  • Auto-return allows user to recall any loaded basket to load door
  • Suggested staining protocols can be easily modified to lab procedures
  • Capable of managing multiple protocols without affecting throughput

Quality Control and Safety Features

  • Displays reagent usage and batch throughput
  • Controls fumes through an easy-to-change carbon filter or an optional external vent adapter kit
  • Offers optional passcode protection to secure customized protocols
  • Battery backup provides approximately 40 min. of power in the event of a power failure
  • Smart software keeps unit's robotic arm from entering user access areas

Physical Specifications

  • D x W x H: Stainer, 28 x 28.5 x 34.5 in. (71 x 72 x 88cm)
  • Power supply, 18.5 x 14 x 5.75 in. (46.3 x 35 x 14.5cm)
  • Weight: Stainer, 176 lb. (80kg); Power supply, 22 lb. (10kg)
  • Electrical requirements: 300VA
  • Two-level design minimizes footprint while making all stations easily accessible

Ordering Information

Available with or without five independently heated positions for drying slides.

Includes:

A total of six water stations, 26 reagent stations, and four configurable loading and unloading stations

  • Charcoal filter
  • Staining basket and carrier (two packs of five each)
  • Three-well water station (two fitted)
  • Pack of six single pot covers
  • Pack of four multi-pot covers
  • 8 ft. (2.5m) long water inlet hose
  • Cytology stainer (without heat)
  • 35 stain pots supplied, fitted
  • Histology stainer (with heat)
  • 30 stain pots supplied, fitted
  • Pack of three spare staining pots
  • Monitor cable
  • Screened data cable
  • Operator manual (specify language)
  • Service record book
Specifications

Specifications

Designed to meet IEC 61010. CSA approved. UL listed. CE marked (IVDD compliant).
34.5 in.
28 in.
20-slide baskets up to 100 slides
28.5 in.
176 lb.
28 x 28.5 x 34.5 in. (71 x 72 x 88cm)
88cm
71cm
Dual voltage
72cm
80kg
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit