Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS290P1

 View more versions of this product

Catalog No. MS290P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Vascular Endothelial Growth Factor (VEGF) Ab-2, Mouse Monoclonal Antibody.

VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. It exhibits potent mitogenic and permeability inducing properties specific for the vascular endothelium. Of the four isoforms of VEGF, the smaller two, VEGF165 and VEGF121, are secreted proteins and act as diffusible agents, whereas the larger two (VEGF189 and VEGF206) remain cell associated.

Host Species: Mouse

Clone: 5C3.F8

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human VEGF from a glioma

Molecular Weight: 19-22kDa (reduced)

Positive Control: VEGF165 recombinant protein

Cellular Localization: Cytoplasmic, cell surface and extracellular matrix Ab-2 does not neutralize the bioactivity of VEGF.

Recommended for:

  • Western Blotting (Reduced only)
  • Neutralization of VEGF (Not suitable)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Vascular Endothelial Growth Factor (VEGF) Ab-2
Monoclonal
200μg/mL
Concentrated
Human VEGF from a glioma
500μL
Angiogenesis
Primary
Human
Western Blot
5C3.F8
Unconjugated
Murine
IgG2a kappa
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit