Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Vasopressin, 98+%

Click to view available options
Quantity:
25mg
100mg
11000-17-2
C46H65N15O12S2
MFCD00076738
Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2; AVP peptide
11000-17-2
C46H65N15O12S2
MFCD00076738
Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Arg-Gly-NH2; AVP peptide
Solubility
Soluble in water (Miscible).

Notes
Store in airtight containers at 2° to 8°. Store in cool, sealed container.
Quantity 100mg
Color White
Solubility Information Soluble in water (Miscible).
Formula Weight 1084.24
Physical Form Lyophilized Powder
Percent Purity ≥98%
Chemical Name or Material Vasopressin

EINECSNumber : 234-236-2

TSCA : No

Recommended Storage : Store at -20°C