Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VASP Mouse anti-Human, Rat, Clone: OTI4D6, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502647S
Description
Vasodilator-stimulated phosphoprotein (VASP) is a member of the Ena-VASP protein family. Ena-VASP family members contain an EHV1 N-terminal domain that binds proteins containing E/DFPPPPXD/E motifs and targets Ena-VASP proteins to focal adhesions. In the mid-region of the protein, family members have a proline-rich domain that binds SH3 and WW domain-containing proteins. Their C-terminal EVH2 domain mediates tetramerization and binds both G and F actin. VASP is associated with filamentous actin formation and likely plays a widespread role in cell adhesion and motility. VASP may also be involved in the intracellular signaling pathways that regulate integrin-extracellular matrix interactions. VASP is regulated by the cyclic nucleotide-dependent kinases PKA and PKG.Specifications
VASP | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P50552 | |
vasodilator stimulated phosphoprotein | |
Mouse | |
IgG1 | |
30 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
361517, 7408 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4D6 | |
Unconjugated | |
VASP | |
Liquid | |
VASP | |
Full length human recombit protein of human VASP produced in 293T Cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human, Rat |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title