Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VASP (p Ser238) Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100822540.025ML
Description
VASP Polyclonal specifically detects VASP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VASP (p Ser238) | |
Polyclonal | |
Unconjugated | |
phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
vasodilator-stimulated phosphoprotein | |
Rabbit | |
50 kDa | |
0.025 mL | |
Phospho Specific | |
7408 | |
Mouse, Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:100 | |
P50552 | |
VASP | |
The antiserum was produced against synthesized phosphopeptide derived from human VASP around the phosphorylation site of serine 238 (K-V-SP-K-Q). | |
Affinity Purified | |
RUO | |
Primary | |
VASP (specific to Phospho-Ser238) detects endogenous levels of VASP only when phosphorylated at serine 238. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only