Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VASP (p Ser238) Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$236.67 - $378.00
Specifications
Antigen | VASP (p Ser238) |
---|---|
Concentration | 1.0 mg/mL |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10082254
|
Novus Biologicals
NB10082254 |
0.05 mL |
Each for $378.00
|
|
NB017890
|
Novus Biologicals
NB100822540.025ML |
0.025 mL |
Each for $236.67
|
|
Description
VASP Polyclonal specifically detects VASP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VASP (p Ser238) | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
P50552 | |
7408 | |
The antiserum was produced against synthesized phosphopeptide derived from human VASP around the phosphorylation site of serine 238 (K-V-SP-K-Q). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Polyclonal | |
Rabbit | |
Phospho Specific | |
vasodilator-stimulated phosphoprotein | |
VASP | |
IgG | |
Affinity Purified | |
50 kDa |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title