Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAT1L Mouse anti-Human, PE, Clone: OTI1H3, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP274831PE
Description
VAT1L Monoclonal antibody specifically detects VAT1L in Human samples. It is validated for Flow Cytometry.Specifications
VAT1L | |
OTI1H3 | |
Flow Cytometry | |
Affinity Purified, Conjugated | |
VAT1L | |
Full length recombinant protein of human VAT1L (NP_065978) produced in HEK293T cells. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Flow Cytometry | |
PE | |
PBS with 0.05% Sodium Azide | |
KIAA1576synaptic vesicle membrane protein VAT-1 homolog-like, vesicle amine transport protein 1 homolog (T. californica)-like, vesicle amine transport protein 1 homolog-like (T. californica) | |
Mouse | |
IgG1 | |
0.1 mL | |
Primary | |
57687.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title