Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAT1L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548513

Catalog No. PA548513


Add to cart

Specifications

Specifications

VAT1L
Polyclonal
PBS with 0.09% sodium azide
Q9HCJ6, Q80TB8
Synaptic vesicle membrane protein VAT-1 homolog-like, 1---, VAT1L, KIAA1576
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
270097, 57687
Western Blot
Unconjugated
VAT1L
Liquid
VAT1L
KLH conjugated synthetic peptide between 391-419 amino acids from the C-terminal region of human VAT1L.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit