Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAT1L Mouse anti-Human, Clone: OTI1H3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525086

Catalog No. PIMA525086


Add to cart

Description

Description

VAT1L is a protein coding gene.
Specifications

Specifications

VAT1L
Monoclonal
0.62 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HCJ6
Synaptic vesicle membrane protein VAT-1 homolog-like, 1---, VAT1L, KIAA1576
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57687
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1H3
Unconjugated
VAT1L
Liquid
VAT1L
Full length human recombit protein of VAT1L produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit