Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vav-1 Mouse anti-Human, DyLight 680, Clone: 557, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243553FR

Catalog No. NBP243553FR


Add to cart

Description

Description

Vav-1 Monoclonal antibody specifically detects Vav-1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Vav-1
557
Western Blot
Purified
VAV1
Recombinant protein encompassing a sequence within the center region of human VAV1. The exact sequence is proprietary.
Protein G purified
Phospho Specific, Signal Transduction
Primary
7409
Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
oncogene vav, proto-oncogene vav, vav 1 guanine nucleotide exchange factor, vav proto-oncogene, VAVvav 1 oncogene
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit