Learn More
VAV1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 163641AP
Catalog No. 50-173-0894
Add to cart
Description
Description
This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14.7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
Specifications
VAV1
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15498
Proto oncogene vav, VAV, VAV1
Rabbit
IgG
150 μL
-20°C
Primary
7409
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
VAV1
Liquid
VAV1
VAV1 Fusion Protein Ag9049
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human