Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VBP1 Mouse anti-Human, DyLight™ 488, Clone: OTI2E6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274836G

Catalog No. NBP274836G


Add to cart

Description

Description

VBP1 Monoclonal antibody specifically detects VBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

VBP1
OTI2E6
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
VBP1
Full length recombinant protein of human VBP1 (NP_003363) produced in HEK293T cells.
Immunogen affinity purified
Core ESC Like Genes, Stem Cell Markers
Primary
7411.0
Immunohistochemistry, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
HIBBJ46, PFD3, PFDN3VHL-binding protein 1, prefoldin 3, prefoldin subunit 3, VBP-1, VHL binding protein-1, von Hippel-Lindau binding protein 1, Von Hippel-Lindau-binding protein 1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit