Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VCAM-1/CD106 Antibody (MVCAM A (429)), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250620FR
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence, Immunocytochemistry.Specifications
| VCAM-1/CD106 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VCAM1 | |
| Stromal cell line PA6 | |
| 0.1 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
| 7412 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence, Immunocytochemistry | |
| MVCAM A (429) | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence, Immunocytochemistry | |
| CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes murine vascular adhesion molecule 1 (VCAM-1), a cell surface glycoprotein that is also known as CD106. CD106 is expressed predominantly on endothelial cells and expression is up-regulated during inflammation. The ligand for CD106 is the alpha 4 subunit (CD49d) of the integrin VLA-4 (CD49d/CD29). Clone MVCAM.A(429) is reported to partially block VCAM-1 mediated functions. | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction