Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCAM-1/CD106 Mouse anti-Human, Canine, Monkey, Biotin, Clone: OTI3H10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274840B

Catalog No. NBP274840B


Add to cart

Description

Description

VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

VCAM-1/CD106
OTI3H10
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
VCAM1
Full length recombinant protein of human VCAM-1/CD106 (NP_001069) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary
7412.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Biotin
PBS with 0.05% Sodium Azide
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Canine, Human, Monkey
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit