Learn More
VCAM-1/CD106 Mouse anti-Human, DyLight 405, Clone: 6G9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP147491V
Catalog No. NBP147491V
Add to cart
Description
Description
VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
VCAM-1/CD106
6G9
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
VCAM1
Purified recombinant fragment of human VCAM-1 expressed in E. coli. [Uniprot: P19320]
Ammonium sulfate precipitation
RUO
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Mouse
IgG1
0.1 ml
Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary
7412