Learn More
VCAM-1/CD106 Rat anti-Mouse, Alexa Fluor 488, Clone: 112734, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB6432G100UG
Catalog No. FAB6432G100
Add to cart
This item is not returnable. View return policy
Description
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
VCAM-1/CD106
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P29533
VCAM1
Mouse myeloma cell line NS0-derived recombinant mouse VCAM-1 Phe25-Glu698 (predicted) Accession # P29533
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects recombinant mouse (rm) VCAM-1 in Western blots and ELISAs. In sandwich immunoassays, no cross-reactivity or interference was observed with recombinant human VCAM-1, rmICAM-1, rmE-Selectin, rmL-Selectin or rmP-Selectin.
Flow Cytometry
112734
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Rat
IgG2a
RUO
Primary
7412
Mouse
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)